Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(methyl methacrylate)

Catalog No. 20033650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-033-650G 50 g
20-033-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-033-650G Supplier Sigma Aldrich Supplier No. 20033650G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly (methyl methacrylate) (PMMA) is an amorphous transparent thermoplastic polymer. PMMA is recognized as an optical polymer based on its refractive index (1.49). Hence, it is used in optical fibers. It finds uses in biological applications because of its lower water absorption capability and biocompatibility. The maximum moisture content of PMMA is 1.71% and SBF absorption is 1.96%. Simulated Body fluid test (SBF) is a method to identify the in vitro bioactivity of ceramic materials, by immersing the materials in an aqueous SBF solution.PMMA may be used as substrate for graphene growth. Some studies report its use asPMMA-titania hybrid optical thins filmsPMMA/polystryrene/clay nanocompositesPMMA/polyurethane/carbon black nanocomposites for methanol fuel cellsacrylic glassimplant in surgical specialtiesas spacer.

Specifications

CAS 9011-14-7
Density 1.200 g/cm3
Linear Formula [CH2C(CH3)(CO2CH3)]n
MDL Number MFCD00134349
Quantity 50 g
Synonym PMMA; Poly(methacrylic acid methyl ester)
Molecular Weight (g/mol) 15000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.