Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cerium(III) chloride heptahydrate

Catalog No. 20298310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-298-310G 10 g
20-298-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-298-310G Supplier Sigma Aldrich Supplier No. 20298310G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cerium chloride heptahydrate is often used to prepare cerium oxide nanoparticles by various methods. It reacts with sodium-oleate to form a cerium-oleate complex which can be further decomposed in hydrocarbon solvents to prepare cerium oxide nanoparticles. Cerium chloride heptahydrate can also be used in the conversion of esters to allylsilanes.

Specifications

CAS 18618-55-8
Quantity 10 g
Molecular Formula CeCl3 -+ 7H2O
Linear Formula CeCl3 -+ 7H2O
MDL Number MFCD00149634
Synonym Cerous chloride heptahydrate
Molecular Weight (g/mol) 372.58
Percent Purity 99.999% trace metals basis
Density ∼3.94 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.