Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver nitrate

Catalog No. 20439050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
2 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-439-050G 50 g
20-439-010G 10 g
20-439-01G 1 g
20-439-0250G 250 g
20-439-02KG 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 20-439-050G Supplier Sigma Aldrich Supplier No. 20439050G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver nitrate

Specifications

Melting Point 212°C
CAS 7761-88-8
Quantity 50 g
Molecular Formula AgNO3
Linear Formula AgNO3
MDL Number MFCD00003414
Synonym Nitric acid silver(I) salt
Molecular Weight (g/mol) 169.87
Percent Purity 99.9999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.