Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium tetrachloropalladate(II)

Catalog No. 20581825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-581-825G 25 g
20-581-81G 1 g
20-581-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-581-825G Supplier Sigma Aldrich Supplier No. 20581825G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tetrachloropalladate(II)[Na2PdCl4] was used to prepared palladium nanospheres, hollow nanopheres and hollow AuxPdY alloy nanoparticles. Na2PdCl4 maybe used in the electroless coating on mild steel and to prepare ion associate cetyltrimethylammonium-tetrachloropalladate which is consequently used to modify carbon based electrodes for better sensitivity.

Specifications

CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Linear Formula Na2PdCl4
Quantity 25 g
MDL Number MFCD00003487
Synonym Palladium(II) sodium chloride
Molecular Weight (g/mol) 294.21
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.