Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium(II) Acetate

Catalog No. 2058691G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
2 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-586-91G 1 g
20-586-92G 2 g
20-586-910G 10 g
20-586-9100G 100 g
20-586-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 20-586-91G Supplier Sigma Aldrich Supplier No. 2058691G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium(II) Acetate

Specifications

CAS 3375-31-3
Melting Point 216.3°C to 223.7°C
Molecular Formula C4H6O4Pd
Linear Formula Pd(OCOCH3)2
MDL Number MFCD00012453
Quantity 1 g
Synonym Pd(OAc)2
Molecular Weight (g/mol) 224.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.