Learn More
Ace Glass, Inc. Lab bottle, Duran, 20L, plain, pouring ring & PTFE lined GL 45PBT cap, cs/1
Supplier: Ace Glass, Inc. 553944
Chemical-resistant and stable 20L Duran laboratory bottles, fabricated from borosilicate glass. Supplied graduated with removable PTFE-lined GL45 PBT cap and pouring ring. Can be hot-air sterilized or autoclaved. Case/1.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.