Learn More
Chemglass Life Sciences Chemglass CG-1924-R-20L 20 L Unjacketed Cylindrical Reaction Vessel, 200 mm Flange
SDP
Supplier:  Chemglass Life Sciences CG1924R20L
Encompass_Preferred
The Chemglass CG-1924-R-20L is a 20 L unjacketed cylindrical reaction vessel engineered for laboratory process chemistry and scale-up. It provides a 200 mm bottom flange, graduated side markings for approximate volume, and a low-dead-space spring-loaded drain valve for efficient, clean drainage and reliable sealing.
  • 20 L capacity.
  • 200 mm flange with o-ring groove for greased seal.
  • Low-dead-space spring-loaded drain valve for flush-bottom drainage.
  • Graduation on both sides for approximate volume measurement.
  • Compatible with 3/4 in compression fitting for valve sidearm.
  • Rated for -60 °C to 230 °C with perfluoro Class VI o-rings.
  • Serial numbered for traceability.
Catalog No. NC3822355