Learn More
JG FINNERAN 20ML CLEAR VIAL 24-400MM 100/C
Supplier:  JG FINNERAN 90882
Encompass_Preferred
Precleaned - 20mL Clear Vial 24-400mm Solid Top White Polypropylene Closure PTFE Lined
Catalog No. NC2308740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.