Learn More
Chemglass Life Sciences Dish, Hexagonal, 9mL, Weighing, Antistatic, Polystyrene
SDP
Supplier:  Chemglass Life Sciences CLS1815007
Encompass_Preferred
Dish, Hexagonal, 9mL, Weighing, Antistatic, Polystyrene. These dishes are excellent for handling solids or liquids during weighing. Dishes are easily bent into pouring spouts that enable non-spill transfer. The molded hexagonal design provides greater balance protection and safety. Not reactive with most substances. Dishes are suitable for weighing of static affected samples. They are stackable for easy storage and accessibility. Dishes will withstand temperatures up to 80°C.
Catalog No. 50-121-5168
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.