Learn More
Marathon LS 20μL LTS-Compatible Pipette Tips, Filtered, Sterile, Low Retention, 96 tips/rack, 10 racks/pack.
SDP
Supplier:  Marathon LS MLSPTFSLTS20
Encompass_Preferred
20μL LTS-Compatible Pipette Tips, Filtered, Sterile, Low Retention, 96 tips/rack, 10 racks/pack. Lot traceable and DNase free, RNase free, and non-pyrogenic. Additional info on MLS-PTFS-LTS20: Made of ultra-clear, medical-grade virgin polypropylene. Molded volume graduation markings. Ultra-low ejection force for comfort, ergonomic pipetting for delivering superior accuracy. Lids have a secure locking tab. Packaged in red rigid autoclavable racks. Filters reduce cross contamination and prevent damaging aerosols from penetrating into the pipette. Racks and grids made from up to 61% recycled materials. MADE IN THE USA. Certifications: ISO13485:2016& ISO9001:2015 These pipette tips are LTS-Compatible and are compatible with Rainin single- and multi-channel LiteTouchpipettors.
Catalog No. NC2609847
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.