Learn More
R&D Systems 20X ANTI-RABBIT SECONDARY IR
SDP
Catalog No. NC2013258 Shop All Novus Products
Encompass
20X Anti-Rabbit Secondary IR conjugate for use on Jess systems ONLY
Catalog No. NC2013258 Supplier R&D Systems Supplier No. 043820/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
20X Anti-Rabbit Secondary IR conjugate for use on Jess systems ONLY