Learn More
Invitrogen™ 20X Bolt™ MES SDS Running Buffer
Description
About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.
Specifications
Specifications
| Chemical Name or Material | Running Buffer |
| Concentration | 20X |
| For Use With (Application) | Protein electrophoresis & western blotting |
| For Use With (Equipment) | Bolt |
| Physical Form | Liquid |
| Product Line | Bolt |
| Quantity | 5 L |
| Formulation | Sodiumdodecyl Sulphate 151-21-3 ( 1-5 ) |
Frequently Asked Questions (FAQs)
DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.
Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.
Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.
The shelf life for Bolt gels is similar to that for NuPAGE gels: 12 months from the date of shipment.
While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.