Learn More
Triforest Enterprises 20X125mm polycarbonate autoclavable culture tube
Supplier:  Triforest Enterprises TPC20125
Encompass_Preferred
TriForest Polycarbonate (PC) culture tubes are optically clear, durable and a safer alternative to glass. They are ideal for production of terminally sterilized products. The tubes are flat bottom and ideal for large scale production culture media, using automated or manual filling systems. PC tubes offer a wide range of chemical resistance as well as excellent strength for repeated use. Leak-proof caps provide security during handling and transportation. TriForest culture tubes maintain excellent pH control. They are provided non sterile with caps seperately packed
Catalog No. NC2401717
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.