Learn More
Sklar Instruments Forceps Jar, Sklar , Stainless Steel, Reusable, Non-Sterile. Sklar Forceps Jars are ideal for holding multiple pairs of forceps and other instruments upright so they can be readily accessed. EA.
Supplier: Sklar Instruments 101541
Forceps Jar, Sklar, Stainless Steel, Reusable, Non-Sterile. Sklar Forceps Jars are ideal for holding multiple pairs of forceps and other instruments upright so they can be readily accessed. The cylandrical shape and uncovered mouth allow handles to rest where they can be easily grasped. The base, of a larger circumference than the rest of the container, ensures the jar does not tip.This jar is measures 2-1/4 in. x 4-1/2 in. with a 3-5/8 in. base. EA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.