Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 21-desacetyldeflazacort-d5 | 13649-57-5 | 99.3% | 404.51 g/mol | C23H24D5NO5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY100085S5MG

Encompass_Preferred

21-Desacetyldeflazacort-d5 is a deuterium-labeled analytical reference standard of 21-desacetyldeflazacort for research use. It is designed to support accurate quantification of the active metabolite in biological samples and to improve precision in method validation and pharmacokinetic studies.

  • Deuterium-labeled isotopologue for mass-spectrometry assays.
  • High purity (>99.3%) for analytical reliability.
  • Molecular weight 404.51 g/mol, well characterized.
  • Suitable as an internal standard for LC-MS/MS and PK studies.
  • Available in small mg quantities for analytical workflows.

Catalog No. 50-002-15426


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.