Learn More
Ellsworth Adhesive Co 21 GAUGE1/2IN PURPLE
Supplier:  Ellsworth Adhesive Co 570587
Encompass
The Loctite 570587 is a straight, 21 gauge, 1/2" long, purple dispensing needle. The needle features a helix thread for secure valve and syringe barrel attachment.
Catalog No. NC2212166
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.