Learn More
Hach Company 2100Q Portable Turbidimeter
Supplier:  Hach Company 2100Q01/DUP
Encompass_Preferred
Measuring and monitoring turbidity is crucial for various industrial and municipalapplications, and Hach’s portable turbidimeter, the 2100Q, provides unsurpassed ease of use and accuracy. With over 80 years of experience, Hach is a leader inturbidity measurement and has consistently developed reliable and durable instruments for customers.  The portable 2100Q turbidity meter is ideal for use in the field or around the plant. It is easy to carry around and take multiple measurements from various collection points in your treatment process. With its intuitive user interface, the 2100Q makes it easy to take measurements and perform calibration and verification. Storing and transferring data from a portable turbidimeter has never been easier.There is also an optional USB+Power module for the 2100Q portable turbidimeter, which allows you to transfer your data directly to a computer. The 2100Q portable turbidimeter is compliant with USEPA Method 180.1 design criteria.
Catalog No. 50-199-7392
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.