Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) perchlorate hexahydrate

Catalog No. 215392100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-539-2100G 100 g
21-539-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-539-2100G Supplier Sigma Aldrich Supplier No. 215392100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The monoclinic structure of copper perchlorate hexahydrate was analyzed by X-ray diffraction. The product may be used as a catalyst during the reaction of 5-methylisophthalaldehyde with 1,3-diaminopropane. Benzaldehyde semicarbazones, reacted with copper perchlorate hexahydrate in acetonitrile, may yield corresponding 1,2,4-triazolin-5-ones.

Specifications

CAS 10294-46-9
Density 2.225 g/mL (at 25°C)
Molecular Formula Cl2CuO8 -+ 6H2O
Linear Formula Cu(ClO4)2 -+ 6H2O
Quantity 100 g
MDL Number MFCD00149666
Synonym Cupric perchlorate hexahydrate
Molecular Weight (g/mol) 370.54
Percent Purity 98%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.