Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-Limonene Oxide, Mixture Of Cis And Trans

Catalog No. 21832450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-832-450G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-832-450G Supplier Sigma Aldrich Supplier No. 21832450G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Limonene oxide, also known as limonene 1,2-epoxide, is a monoterpene that can be prepared fro mLimonene. It has antinociceptive and antitumoral activities and is used in many products like food additives, cosmetics, and perfumes.(+)-Limonene oxide (mixture of cis and trans) undergoes biocatalytic resolution in the presence of epoxide hydrolase to form enantiomerically pure cis- and trans-limonene oxides.

Specifications

CAS 26348-61-8
Melting Point 130°C to 132°C
Molecular Formula C5H11NO3 -+ HCl
Linear Formula HOCH2CH(NH2)CO2C2H5-+HCl
Refractive Index n20/D 1.466
Quantity 5 g
MDL Number MFCD00012594
Molecular Weight (g/mol) 169.61 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.