Learn More
Fisherbrand™ Safety Blood Collection Needles
Catalog No. 22024242 Shop All Fisherbrand Products
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Preattached Holder:
No
Yes
Product Type:
Safety Blood Collection Needle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Preattached Holder Product Type
22-024-242 25 G No Safety Blood Collection Needle
22-024-240 21 G No Safety Blood Collection Needle
22-024-238 23 G Yes Safety Blood Collection Needle
22-024-241 23 G No Safety Blood Collection Needle
22-024-237 21 G Yes Safety Blood Collection Needle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-024-242
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fisherbrand Safety Blood Collection Needles are safety-engineered devices intended for venous blood collection. Needles with the preattached holder safety mechanism can be activated immediately after blood draw, helping to protect you against needlestick injury.
  • Designed to help reduce the risk of needlestick injury
  • Thin walled needle to minimize patient discomfort
Specifications
Product Type Safety Blood Collection Needle
Needle Gauge 25 G
Sterility Sterile
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.
Preattached Holder No