Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Autoclavable HD Biohazard Waste Bags
SDP

Catalog No. 22044565 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear with Red Print
Red with Black Print
Dimensions (L x W):
12 x 8 in. (30 x 20 cm)
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
35 x 25 in. (89 x 64 cm)
36 x 24 in. (91 x 61 cm)
36 x 45 in. (91 x 114 cm)
Thickness:
1.25 mil
1.50 mil
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W) Thickness
22-044565 Red with Black Print 36 x 45 in. (91 x 114 cm) 1.50 mil
22-044559 Clear with Red Print 30 x 24 in. (76 x 61 cm) 1.50 mil
22-04-4558 Clear with Red Print 24 x 12 in. (61 x 30 cm) 1.50 mil
22-044561 Red with Black Print 12 x 8 in. (30 x 20 cm) 1.25 mil
22-044562 Red with Black Print 19 x 14 in. (48 x 36 cm) 1.25 mil
22-044560 Clear with Red Print 36 x 24 in. (91 x 61 cm) 1.50 mil
22-044563 Red with Black Print 23 x 19 in. (58 x 48 cm) 1.25 mil
22-044564 Red with Black Print 35 x 25 in. (89 x 64 cm) 1.50 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22-044565
Add to Cart
Add to Cart

Strong and leak-resistant

  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure

Specifications

Closure Type Twist Tie
Color Red with Black Print
Dimensions (L x W) 36 x 45 in. (91 x 114 cm)
Length (Metric) 114 cm
Width (Metric) 91 cm
Autoclavable Autoclavable
Includes Biohazard symbol and text in English and Spanish
Width (English) 45 in.
Length (English) 36 in.
Thickness 1.50 mil
Material Polyethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.