Learn More
StatLab KoH Fungal Control Vial
SDP
Catalog No. 22130493
Change view
Click to view available options
Product Type:
Fungal Control
Catalog No. Product Type
22-130-493 Fungal Control
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-493 Supplier StatLab Supplier No. 6025
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Harmful if swallowed, in contact with skin, or if inhaled
An ideal control for skin scrapings
  • The 1 mL vial is filled with inactivated mycelia and spores of Aspergillis sp., along with cellular debris, for quick characterization of specimens
  • An addition of Methylene blue can be used to help highlight the mycelia
  • 1 cc vial
Specifications
Product Type Fungal Control
Sterility Non-sterile
Quantity 1 mL
Species Aspergillus species