Learn More
StatLab Modified Auramine O Set - TB Rapid Stain Kit

Description
- Organisms are visible at low power for fast scanning and produce little to no background fluorescence
- Includes one stain and one decolorizer
- Contains 4 bottles of stain, and 4 bottles of decolorizer/quencher
- Available with either 1 L or 250 mL bottles
Specifications
Specifications
| Quantity | 4 Bottles Stain, 4 Bottles Decolorizer |
| Volume (Metric) | 1 L Stain, 1 L Decolorizer |
| Product Type | Auramine O |
| Packaging | 250 L Bottles |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.