Learn More
Fisher Healthcare™ Pinnacle Portfolio Hematoxylin 1 Stain
Catalog No. 22220101 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 x 500 mL
Catalog No. Quantity
22-220-101 4 x 500 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-220-101
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapid stain requires no filtration prior to use
Hematoxylin 1 Stain Features:
  • Produces staining intensities similar to Gill-1, -2, and -3 Hematoxylins, based on timing
  • Will not form crystals upon standing or exposure to cold weather
  • Developed to prevent the formation of metallic sheens
  • Does not contain alcohol
Pinnacle Portfolio
  • Designed as a system
  • Each component has added value and benefits that reduce cost per slide
  • Unique chemical formulae of the Pinnacle Portfolio promote long-term chemical stability, which extends the usable life of each component
  • Subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity
Differentiation is not necessary; however, Clarifier™ 1 should be used to eliminate any background staining caused by the use of excessive water bath adhesive
Specifications
Product Type Hematoxylin
Quantity 4 x 500 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov