Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzylmagnesium Chloride Solution

Catalog No. 225916800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-591-6800ML 800 mL
22-591-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-591-6800ML Supplier Sigma Aldrich Supplier No. 225916800ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzylmagnesium chloride (Grignard reagent) can be used as an alkylating reagent:For alkylating quinolyl-functionalized Cp-chromium(III) complexes which are used as precursors for the preparation of active olefin polymerization catalysts. In the synthesis of 2,3-disubstituted benzopyran-4-ones, 6-chloro-8-methylpurine derivative, and phenylalanine derivatives.

Specifications

CAS 6921-34-2
Density 1.031 g/mL at 25°C
Molecular Formula C7H7ClMg
Linear Formula C6H5CH2MgCl
Quantity 800 mL
MDL Number MFCD00000469
Molecular Weight (g/mol) 150.89 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.