Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC #226 Platinum cured, class VI ,Silicone O ring
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC 10191289
Encompass_Preferred
This item has a minimum order quantity of 50 per supplier requirements.
#226 Platinum cured, class VI,silicone o ring, single pack.Medical grade doublebagged, peel away pouch.Cleaned and packaged in ISOclass 7 cleanroom. Gammairradiated, 25-40 kGy
Catalog No. NC1717377
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.