Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt(II) sulfate hydrate

Catalog No. 22959810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-959-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-959-810G Supplier Sigma Aldrich Supplier No. 22959810G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cobalt(II) sulfate hydrate (CoSO4.H2O) can be used in the synthesis of coordination polymers. It can also be used as a starting material in the synthesis of cobalt sulfides hexagonal nanocrystallites.

Specifications

CAS 60459-08-7
Density 2.03 g/mL (at 25°C)
Molecular Formula CoO4S -+ xH2O
Linear Formula CoSO4 -+ xH2O
MDL Number MFCD00149657
Quantity 10 g
Synonym Cobaltous sulfate
Molecular Weight (g/mol) 155
Percent Purity 99.998% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.