Learn More
Justrite™ 23 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Manual-Closing Doors
Shop All Justrite Safety Group Products
Description
- Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
- Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
- Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
- Polyethylene accessories with a slightly curved edge provide additional protection against leaks and spills
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Load Bearing Capacity | 23 gal. |
| Door Style | Manual-close |
| Color | Silver |