Learn More
DYNAMIC DEVICES Lynx Series 96 Tip Pipette Head, 230uL, 4800/case (50 racks of 96 tips)
Supplier:  DYNAMIC DEVICES DDX96230F1
Encompass
Case includes 4,800 total tips packaged in 50 racks of 96 tips in a robotic box. 230 μL (Discount pricing available if ordering 2-4 cases)
Catalog No. NC2037796