Learn More
Fisher Healthcare™ PROTOCOL™ Hema-Quik™ and Hema-Quik II Stain Solutions
Shop All Fisherbrand Products

Description
Stains for manual and automated staining methods.
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Remain stable for a minimum of 12 months when stored at room temperature
- Allow varied staining times to suit individual preferences
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 1 L |
| Storage Requirements | Room Temperature |
| Boiling Point | 65°C |
| Format | Liquid |
| Color | Dark Blue |
| Product Type | Hema-Quik II Stain Solution |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.