Learn More
QuidelOrtho Bovine Albumin for Blood Typing Reactions
SDP
Catalog No. 23287246
Change view
Click to view available options
Quantity:
15 x 10 mL
100 x 10 mL
Type:
Bovine Serum Albumin Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-287246 15 x 10 mL Bovine Serum Albumin Solution
23-287-456 100 x 10 mL Bovine Serum Albumin Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-287246 Supplier QuidelOrtho Supplier No. 718280
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Enhances antibody reactions
  • Reagents are used to enhance antigen/antibody interactions
  • Results in an increase in speed and sensitivity of detection
Specifications
Type Bovine Serum Albumin Solution
For Use With (Application) For Blood typing reactions, enhance Antigen/antibody interactions, increase in speed and sensitivity of detection
Quantity 15 x 10 mL