Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 234X Uncoated HPPE Cut-Resistant Gloves

Catalog No. 19021009 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
2X-Large, 10
Large, 8
Medium, 7
Small, 6
X-Large, 9
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-021-009 2X-Large, 10
19-021-005 Small, 6
19-021-006 Medium, 7
19-021-007 Large, 8
19-021-008 X-Large, 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-021-009 Supplier SHOWA™ Supplier No. 234X10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use in 'knife hand' applications in food processing, these flexible cut protection gloves feature a HPPE liner for protection in both dry and greasy environments.

  • Resilient cut-resistant glove offering comfort and flexibility
  • ANSI cut level A4 and EN 388:2016 level D
  • Cooling HPPE properties and breathable back of hand reduces perspiration and keeps hands dry
  • Designed for use in knife hand applications in food processing and food service industries
  • Can be used as a glove liner for added protection and comfort
  • Thin and lightweight for enhanced dexterity and longer use
  • Launderable for re-use, leads to less waste and cost efficiency
  • Seamless knit designed to prevent irritation

Specifications

Certifications/Compliance FDA Compliant, CE Cat. 2
Certifications/Compliance (Europe) Cat II CE, EN 407
Construction Seamless Knit
Coating Material Uncoated
Color Gray
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Latex Free No
Material HPPE, Engineered Yarn
Product Type Cut-resistant Gloves
Size 2X-Large, 10
EN388 Standard EN 388:1X41D
For Use With (Application) Beverage and food packing and handling
Glove Cuff Color White
Liner Material HPPE
Model 234X-10
Puncture Rating ASTM F2878 LEVEL 0
Series 234X Series
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.