Learn More
CLICKBIO INC XBLOK 2412 E-Beamed Irradiated (20/CS)
Supplier:  CLICKBIO INC XBLOK24121E
Encompass
The XBLOK2412 is a polypropylene ANSI-SLAS footprint plate with 24 x 12 mL wells. It is ideal for parallel fermentation and sample preparation workflows. It is e-beam irradiated for aseptic workflows.
Catalog No. NC1914746
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.