Learn More
Medchemexpress LLC HY-W007584 500mg Medchemexpress, 2,4-Dimethoxybenzyl alcohol CAS:7314-44-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW007584500MG
Encompass_Preferred
Medchemexpress, HY-W007584 500mg 2,4-Dimethoxybenzyl alcohol CAS:7314-44-5 2,4-Dimethoxybenzyl alcohol, an aromatic alcohol, is a substrate of glucose-methanol-choline (GMC) oxidoreductase. GMC oxidoreductase displays the characteristics of an aryl-alcohol oxidase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3243
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.