Learn More
Thermo Scientific™ Oxoid™ 24 LEB Complete
Catalog No. CM1154R Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Each
10 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CM1154R 2.5 kg
CM1154B 1 Each
CM1154G 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. CM1154R Supplier Thermo Scientific™ Supplier No. CM1154R
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
24 LEB complete is used for the enrichment of Listeria sp and is pre-supplemented with 24 LEB Selective Supplement (does not require the addition of SR0243E)
Selectively enrichment of Listeria monocytogenes and other Listeria spp. from food and environmental samples with Thermo Scientific™ 24 Listeria Enrichment Broth (24 LEB) (Dehydrated). The medium provides optimal growing conditions for all Listeria species for PCR applications. It is specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria.
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. 24 Listeria Enrichment Broth (24 LEB) is designed to identify Listeria spp. based on their utilization of a chomogenic substrate. Easy-to-read: chromogenic and phospholipase enzyme reactions easily differentiate Listeria monocytogenes from other Listeria spp.Adheres to ISO 11290-1:1997 standards.The presence of the chromogenic substrate for the detection of glucosidase (an enzyme common to all Listeria spp. and to a few strains of Enterococci and Bacilli) results in chromogenic activity. Further differentiation of Listeria monocytogenes and other Listeria spp. is facilitated by the addition of phosphatidylinositol or lethicin substrate for the detection of phospholipase enzyme.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microbiology Media
For Use With (Application) PCR applications
For Use With (Equipment) Thermo Scientific™ Oxoid 24 Listeria enrichment broth
Quantity 2.5 kg