Learn More
Thermo Scientific™ 24-Place Fixed-Angle Rotors with Covers
Catalog No. 75007599
Change view
Click to view available options
Max. Speed:
13,000 rpm
15,000 rpm
Max. RCF:
16,060 x g
21,890 x g
Angle:
40°
45°
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. Speed Max. RCF Angle
75-007-599 13,000 rpm 16,060 x g 40°
75-007-593 15,000 rpm 21,890 x g 45°
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-007-599 Supplier Thermo Scientific™ Supplier No. 75007599