Learn More
Organomation 24-POS N-EVAP 120V 900W
SDP
Supplier:  Organomation 11250CB
Encompass_Preferred
• Stainless steel water bath; 120 V; 900 W• Electronic digital temperature control box• Pressure regulator and timer for heat/gas flow• Nitrogen distribution manifold• Gas flow meter; 0-20 L/min• Two dozen stainless steel needles; 4 in x 19 ga• For glassware 10-30 mm in diameter
Catalog No. NC2406551
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.