Learn More
Enzo Life Sciences 24(S)-Hydroxycholesterol (5mg). CAS: 474-73-7
SDP
Supplier:  Enzo Life Sciences BMLGR2300005
Encompass_Preferred
Endogenous agonist for the nuclear receptor LXR. Important metabolite of cholesterol in the brain. Displays neurotoxic effects on human neuroblastoma cells. Cerebrospinal fluid concentrations of 24(S)-hydroxycholesterol are increased in patients with Alzheimer’s disease and mild cognitive impairment. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (10mg/ml) or DMSO (10mg/ml - requires warming to ~40°C). Long Term Storage: -20°C.
Catalog No. 50-201-0936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.