Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 24(S)-Hydroxycholesterol, Endogenous LXR Agonist, >98% Purity, 5 mg

Catalog No. 502706492
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6492 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6492 Supplier Abcam Supplier No. AB1416325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endogenous LXR agonist involved in cholesterol homeostasis and induction of necroptosis.

  • Molecular weight: 402.7 Da
  • Purity: >98%
  • Biological activity: Endogenous agonist for LXR (EC50 values are 4 μM and 3 μM at LXRα and LXRβ respectively)
  • Function: Involved in cholesterol homeostasis; induces neuronal cell death by necroptosis
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Shipping Condition Ambient
Target Molecule LXR (LXRα, LXRβ)
Quantity 5 mg
For Use With (Application) Cholesterol homeostasis research, LXR agonist studies, Necroptosis research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.