Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ QuickPrep™ 2 Sample Holder

Catalog No. MP116005512 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Volume (Metric):
24 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
MP116005512 24 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116005512 Supplier MP Biomedicals™ Supplier No. 116005512
Add to Cart
Add to Cart

Compatible with the QuickPrep System. QuickPrep™ 2 Sample Holder is a 24 x 2 mL QuickPrep sample tube holder for room temperature lysis and grinding on FastPrep-24.

  • Adapter for FastPrep-24 Instruments to grind, lyse, homogenize 24 x 2 mL samples.
  • The QuickPrep adapter is ideal for high-throughput applications and can process up to 24 x 2 mL samples simultaneously.
  • It is the standard adapter supplied with the FastPrep-24™ 5G benchtop instrument.
  • The ergonomic design of the adapter ensures the 2 mL tubes are easy-to-load and they remain securely sealed during grinding, lysing and homogenization.
  • It is stable and can be placed on a workspace to be used as a tube rack.
  • Frozen samples loaded in the adapter and stored at -20°C to -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
  • Purchase multiple adapters to use the same FastPrep-24 5G instrument for multiple lab groups or sample applications.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 24 x 2 mL
Storage Requirements Room Temperature (15°C to 30°C)
Quantity 1 Ea.
For Use With (Equipment) FastPrep-24
Product Type Tube Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.