Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Quinoline

Catalog No. 2415715G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-157-15G 5 g
24-157-1100G 100 g
24-157-1500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24-157-15G Supplier Sigma Aldrich Supplier No. 2415715G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Quinoline

Specifications

CAS 91-22-5
Melting Point -17°C to +13°C
Density 1.093 g/mL (at 25°C)
Boiling Point 113°C to 114°C (11 mmHg); 237°C
Molecular Formula C9H7N
Linear Formula C9H7N
Refractive Index n20/D 1.625 (literature)
MDL Number MFCD00006736
Quantity 5 g
Synonym 1-Benzazine; 2,3-Benzopyridine
Molecular Weight (g/mol) 129.16
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.