Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Iodo-2-methylpropane

Catalog No. 244732100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-473-2100G 100 g
24-473-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-473-2100G Supplier Sigma Aldrich Supplier No. 244732100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The liquid and solid-state infrared spectra of 1-iodo-2-methylpropane were studied.

Specifications

CAS 513-38-2
Density 1.599 g/mL (at 25°C (literature))
Boiling Point 120°C to 121°C (lit.)
Molecular Formula C4H9I
Refractive Index n20/D 1.496 (literature)
Linear Formula (CH3)2 CHCH2I
MDL Number MFCD00001084
Quantity 100 g
Synonym Isobutyl iodide
Molecular Weight (g/mol) 184.02
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.