Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-N-(Trifluoroacetyl)Pyrrolidine-2-Carbonyl Chloride Solution

Catalog No. 2485095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-850-95G 5 g
24-850-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 24-850-95G Supplier Sigma Aldrich Supplier No. 2485095G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-N-(Trifluoroacetyl)pyrrolidine-2-carbonyl chloride can be used as a chiral derivatization reagent:In the chiral separation of psychoactive, cathinone- and amphetamine-related drugsusing GC-MS technique.In the estimation of cathinone related drug enantiomers in biological samples like urine and plasma using GC-MS technique.

Specifications

CAS 36724-68-2
Density 1.308 g/mL at 25°C
Molecular Formula C7H7ClF3NO2
Refractive Index n20/D 1.424
Linear Formula C7H7ClF3NO2
MDL Number MFCD00010418
Quantity 5 g
Synonym (S)-(-)-N-(Trifluoroacetyl)prolyl chloride solution; TPC
Molecular Weight (g/mol) 229.58 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.