Learn More
Beckman Coulter Open Top Tubes, 358125
Supplier:  Beckman Coulter 358125
Encompass
The Beckman Coulter 358125 is a 15mL polypropylene conical centrifuge tube with a flat-top screw cap. It has graduated markings up to 15mL and can withstand centrifugal forces up to 5,000 x g. The tubes are made from clear polypropylene for visibility of contents. They can be autoclaved and are non-pyrogenic and non-cytotoxic. The tubes have wide openings for easy sample transfer and are suitable for a variety of laboratory applications including sample preparation, storage, and centrifugation. The tubes come sterile and individually wrapped.
Catalog No. NC2175704
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.