Learn More
Seracare Life Sciences Inc 25% Human Serum Albumin Diagnostic Grade, Liquid, 100mL
Supplier:  Seracare Life Sciences Inc 18600025
Encompass
SeraCare's 25% Human Serum Albumin diagnostic grade solution is manufactured from human plasma tested per FDA guidelines. The plasma is negative for HBsAg, anti-HIV 1/2, anti-HCV, HIV-1 RNA, HCV RNA and HBV DNA. Each donor was tested and found negative for syphilis per FDA guidelines. SeraCare's Human Serum Albumin solution offers high purity, low bioburden and is suitable for assay design, life science, and cell culture applications.
Catalog No. NC1658529
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.