Learn More
Enzo Life Sciences 25-Hydroxyvitamin D3 (5mg). CAS: 19356-17-3
SDP
Supplier:  Enzo Life Sciences BMLDM1000005
Encompass_Preferred
Intermediate in the biosynthesis of 1α,25-dihydroxyvitamin D3. Major circulating metabolite of vitamin D3. Alternative name: Calcifediol, 25-Hydroxycholecalciferol. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0514
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.