Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ 25 mL Glass Burette, Straight Bore, PTFE Stopcock, Sure Trace Certification
BIOPHARMA

Catalog No. 12183069 Shop All Eisco Products
Change view
Click to view available options
Scale:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Scale
12-183-069 Blue
12-183-068 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-183-069 Supplier Eisco™ Supplier No. CH0240BST

missing translation for 'validMainframeMsg'

Burette, 25 mL, 0.10 mL Graduations, Class A, ± 0.05 mL Tolerance. Borosilicate Glass with PTFE Key Stopcock.

  • 25 mL Burette with 0.1 mL graduations
  • Class A, ± 0.05 mL tolerance as per ASTM DIN ISO 385 standards for precision
  • Includes an attached PTFE straight bore stopcock
  • Burette is made of precisely bore high quality, heavy duty Borosilicate 3.3 glass
  • Features chemical resistant printing
  • All contents are included in a single package

Specifications

Capacity (Metric) 25 mL
Length (Metric) 60.96 cm
Material Borosilicate Glass (3.3)
Graduations 0.1 mL
Scale Blue
Type Buret
Class Class A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.