Learn More
Genscript Corporation cTnI 5C7 mAb Mouse
Supplier:  Genscript Corporation V0030110
Encompass_Preferred
Cardiac Troponin I (cTnI) is synthesized in cardiac muscle and forms a structural complex with Troponin T and C Its concentration in blood increases after the onset of chest pain and maintains peak for a couple of weeks The cTnI protein is considered as useful diagnosis marker for acute myocardial infarction (AMI)
Catalog No. 50-000-58521
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.