Learn More
Across International 250C UL Certified 3.2 CF Vacuum Oven with 3 Heating Shelves
SDP
Supplier:  Across International AT32UL.110
Encompass_Preferred
Introducing the new UL certified 3.2. cu ft vacuum ovens, this certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan. Also come standard is the secondary over-temp protection dial, stainless steel tubing, stainless steel vacuum & vent valve, oil-filled vacuum gauge, and two year warranty. With our new internal shelf heating technology, each shelf in these ovens has its own heater, in-shelf temperature sensor and temperature controller, together with great thermal-conductive aluminum and our new low proportional controller technologies, the result is perfect uniformity, accurate temperature, super-fast heating rates, minimum heat loss/exterior temperature and very low power consumption.
Catalog No. 50-225-1189
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.