Learn More
Chemglass Life Sciences 250mL Filtering Flask, Heavy Wall, 24/40 Outer Joint, Hose Connection
SDP
Supplier:  Chemglass Life Sciences CG155001
Encompass_Preferred
250mL Filtering Flask, Heavy Wall, 24/40 Outer Joint, Hose Connection. HEAVY WALL Erlenmeyer shaped filtering flask with standard taper outer joint and serrated hose connection for attachment to vacuum source. For stoppers see CG-3000.
Catalog No. CG155001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.